Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052905.3 | NP_443137.2 | 20 | 2865 | ||
XM_005246263.2 | XP_005246320.1 | 20 | 2932 | |||
XM_005246265.3 | XP_005246322.1 | 20 | 2932 | |||
XM_011510530.1 | XP_011508832.1 | 21 | 2953 | |||
XM_011510531.1 | XP_011508833.1 | 21 | 2953 | |||
XM_011510532.2 | XP_011508834.1 | 21 | 2953 | |||
XM_011510533.2 | XP_011508835.1 | 21 | 2953 | |||
XM_011510534.1 | XP_011508836.1 | 20 | 2884 | |||
XM_011510535.2 | XP_011508837.1 | 20 | 2800 | |||
XM_011510536.2 | XP_011508838.1 | 15 | 1904 | |||
GenBank mRNA | AB067489.1 | - | 20 | 2636 | ||
AK094865.1 | - | 4 | 420 | |||
AK131316.1 | - | 6 | 1063 | |||
AL834396.1 | - | 18 | 2249 | |||
BC113878.1 | - | 7 | 827 | |||
BC114438.1 | - | 7 | 832 | |||
BC167159.1 | - | 20 | 2636 | 8862581 |