Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085487.2 | NP_001078956.1 | 15 | 1845 | ||
XM_006710314.3 | XP_006710377.1 | 15 | 1861 | |||
XM_011540573.2 | XP_011538875.1 | 16 | 1825 | |||
XM_011540574.2 | XP_011538876.1 | 15 | 2093 | |||
XM_017000195.1 | XP_016855684.1 | 16 | 1897 | |||
XM_017000196.1 | XP_016855685.1 | 13 | 1747 | |||
XM_017000197.1 | XP_016855686.1 | 11 | 1525 | |||
GenBank mRNA | AB067502.1 | - | 11 | 1871 | ||
AK126835.1 | - | 9 | 1394 | |||
AK292919.1 | - | 2 | 532 | |||
BX537912.1 | - | 2 | 109 | |||
BX648168.1 | - | 12 | 1667 |