Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301627.1 | NP_001288556.1 | 8 | 1311 | ||
NM_052917.3 | NP_443149.2 | 10 | 1789 | |||
XM_011510537.2 | XP_011508839.1 | 10 | 1784 | |||
XM_011510538.2 | XP_011508840.1 | 8 | 1196 | |||
XM_017003258.1 | XP_016858747.1 | 10 | 1638 | |||
XM_017003259.1 | XP_016858748.1 | 9 | 1363 | |||
XM_017003260.1 | XP_016858749.1 | 10 | 1784 | |||
XM_017003261.1 | XP_016858750.1 | 10 | 1648 | |||
XM_017003262.1 | XP_016858751.1 | 8 | 1054 | |||
XM_017003263.1 | XP_016858752.1 | 8 | 1196 | |||
GenBank mRNA | AB067505.1 | - | 8 | 1089 | ||
AB078142.1 | - | 8 | 1207 | |||
AJ505991.2 | - | 8 | 1207 | |||
AK123152.1 | - | 1472 | ||||
AK131195.1 | - | 10 | 1734 | |||
AK308675.1 | - | 8 | 1196 | |||
BC101031.1 | - | 8 | 1207 | 40007710 | ||
BC101032.2 | - | 8 | 1207 | 40007711 | ||
BC101033.1 | - | 8 | 1207 | 40007712 | ||
BC101034.2 | - | 8 | 1207 | 40007713 | ||
KJ534843.1 | - | 8 | 1207 | |||
KJ535058.1 | - | 7 | 1038 |