Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242508.1 | NP_001229437.1 | 12 | 1294 | ||
NM_018030.4 | NP_060500.3 | 10 | 1663 | |||
NM_080597.3 | NP_542164.2 | 24 | 2550 | |||
XM_006722380.3 | XP_006722443.1 | 23 | 2463 | |||
XM_006722382.3 | XP_006722445.1 | 23 | 2428 | |||
XM_017025530.1 | XP_016881019.1 | 24 | 2842 | |||
XM_017025531.1 | XP_016881020.1 | 22 | 2152 | |||
XM_017025533.1 | XP_016881022.1 | 10 | 1146 | |||
XM_017025534.1 | XP_016881023.1 | 10 | 3674 | |||
GenBank mRNA | AF274714.1 | - | 10 | 1663 | ||
AF323726.2 | - | 24 | 2438 | |||
AF392449.1 | - | 10 | 1284 | |||
AF392450.1 | - | 24 | 2507 | |||
AK001079.1 | - | 11 | 1191 | |||
AK096358.1 | - | 24 | 2550 | |||
AK292895.1 | - | 25 | 2482 | |||
AK301862.1 | - | 12 | 1294 | |||
AK314552.1 | - | 10 | 1114 | |||
BC063420.1 | - | 10 | 1349 | |||
BC073854.1 | - | 8 | 870 | |||
BC098384.1 | - | 10 | 1342 | |||
BC136452.1 | - | 24 | 2447 | 9020441 | ||
BC144117.1 | - | 24 | 2447 | 9052637 |