Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145798.2 | NP_665741.1 | 22 | 2583 | ||
XM_017024122.1 | XP_016879611.1 | 22 | 2605 | |||
XM_017024123.1 | XP_016879612.1 | 22 | 2723 | |||
XM_017024124.1 | XP_016879613.1 | 21 | 2570 | |||
XM_017024125.1 | XP_016879614.1 | 12 | 1506 | |||
GenBank mRNA | AB208886.1 | - | 10 | 3430 | ||
AF323729.1 | - | 22 | 2572 | |||
AF392446.1 | - | 22 | 2636 | |||
BC028010.1 | - | 14 | 1771 | |||
BC065482.1 | - | 22 | 2555 | 6081784 |