Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030968.3 | NP_112230.1 | 4 | 932 | ||
NM_153372.1 | NP_699203.1 | 4 | 668 | |||
NM_198593.3 | NP_940995.1 | 4 | 748 | |||
NM_198594.2 | NP_940996.1 | 3 | 763 | |||
NR_049769.1 | 4 | 786 | ||||
XM_006721663.1 | XP_006721726.1 | 3 | 793 | |||
XM_006721664.1 | XP_006721727.1 | 6 | 1380 | |||
XM_006721666.1 | XP_006721729.1 | 5 | 996 | |||
XM_017024126.1 | XP_016879615.1 | 3 | 1037 | |||
GenBank mRNA | AF232905.1 | - | 3 | 875 | ||
AF329840.1 | - | 4 | 669 | |||
AJ272138.1 | - | 3 | 875 | |||
AK055541.1 | - | 3 | 578 | |||
AK075175.1 | - | 2 | 273 | |||
AK172767.1 | - | 4 | 769 | |||
AK292034.1 | - | 4 | 786 | |||
AY358424.1 | - | 4 | 898 | |||
BC021553.1 | - | 4 | 554 | 4622952 |