Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031908.4 | NP_114114.2 | 3 | 645 | ||
XM_011534425.2 | XP_011532727.1 | 4 | 777 | |||
XM_011534426.2 | XP_011532728.1 | 5 | 573 | |||
XM_011534427.2 | XP_011532729.1 | 5 | 668 | |||
XM_017009013.1 | XP_016864502.1 | 4 | 456 | |||
GenBank mRNA | AF329836.1 | - | 3 | 653 | ||
AY358839.1 | - | 3 | 645 | |||
BC011699.2 | - | 3 | 626 | |||
BC054506.1 | - | 3 | 619 |