Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052932.2 | NP_443164.2 | 3, 4 | 862 | ||
GenBank mRNA | AK075420.1 | - | 3, 4 | 646 | ||
AK225790.1 | - | 3, 4 | 833 | |||
AY008283.1 | - | 2, 3 | 600 | |||
AY157580.1 | - | 3, 4 | 557 | |||
AY358696.1 | - | 3, 4 | 534 | |||
BC032296.1 | - | 3, 4 | 549 | 4340763 |