Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004051.3 | NP_001004051.1 | 5 | 3247 | ||
NM_001184874.2 | NP_001171803.1 | 5 | 3375 | |||
NM_001184875.2 | NP_001171804.1 | 4 | 3287 | |||
NM_001184876.2 | NP_001171805.1 | 5 | 3286 | |||
NM_138437.5 | NP_612446.1 | 4 | 3159 | |||
GenBank mRNA | AK092981.1 | - | 1 | 335 | ||
AK094646.1 | - | 2 | 1545 | |||
AK096067.1 | - | 1 | 2496 | |||
AK123832.1 | - | 4 | 3159 | |||
AL832187.1 | - | 1 | 1887 | |||
BC013576.1 | - | 1 | 2491 | 3893954 | ||
BC036772.1 | - | 4 | 2923 | 5723418 | ||
BC051707.1 | - | 4 | 3637 |
Gene Symbol: ARMCX5-GPRASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199818.1 | NP_001186747.1 | 7 | 3379 |