Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004051.3 | NP_001004051.1 | 5 | 2010 | ||
NM_001184874.2 | NP_001171803.1 | 5 | 2138 | |||
NM_001184875.2 | NP_001171804.1 | 4 | 2050 | |||
NM_001184876.2 | NP_001171805.1 | 5 | 2049 | |||
NM_138437.5 | NP_612446.1 | 4 | 1922 | |||
GenBank mRNA | AK094646.1 | - | 1 | 370 | ||
AK096067.1 | - | 1 | 1258 | |||
AK123832.1 | - | 4 | 1922 | |||
AL832187.1 | - | 1 | 650 | |||
BC013576.1 | - | 1 | 1254 | 3893954 | ||
BC036772.1 | - | 4 | 1686 | 5723418 | ||
BC051707.1 | - | 4 | 2400 |
Gene Symbol: ARMCX5-GPRASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199818.1 | NP_001186747.1 | 7 | 2142 |