Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006615.2 | NP_001006616.1 | 9 | 973 | ||
NM_001012361.3 | NP_001012361.1 | 10 | 1106 | |||
NM_145241.4 | NP_660284.1 | 10 | 1103 | |||
NR_134677.1 | 10 | 1074 | ||||
XM_011518194.2 | XP_011516496.1 | 9 | 1017 | |||
GenBank mRNA | AK074891.1 | - | 9 | 887 | ||
AK093240.1 | - | 10 | 1035 | |||
AK313843.1 | - | 10 | 1045 | |||
AL833635.1 | - | 10 | 991 | |||
BC012352.1 | - | 10 | 1035 | 4634445 | ||
BC022982.1 | - | 10 | 1032 |