Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MB21D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138441.2 | NP_612450.2 | 4 | 1280 | ||
XM_017010232.1 | XP_016865721.1 | 4 | 1290 | |||
GenBank mRNA | AK097148.1 | - | 4 | 1236 | ||
BC012928.1 | - | 4 | 1219 | |||
BC108714.1 | - | 4 | 1239 | 6015929 | ||
BC113606.1 | - | 4 | 1216 | 8322658 | ||
BC113608.1 | - | 4 | 1216 | 8322660 | ||
BC143694.1 | - | 4 | 1216 | 9052208 | ||
KC294566.1 | - | 4 | 1141 |