Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052935.4 | NP_443167.4 | 7 | 579 | ||
NR_033464.1 | 7 | 647 | ||||
NR_033465.1 | 6 | 803 | ||||
XM_006721669.3 | XP_006721732.1 | 8 | 772 | |||
XM_006721670.3 | XP_006721733.1 | 8 | 663 | |||
XM_011524276.2 | XP_011522578.1 | 7 | 584 | |||
XM_011524277.2 | XP_011522579.1 | 7 | 656 | |||
GenBank mRNA | BC013742.1 | - | 6 | 624 | 4470687 | |
BC014132.2 | - | 6 | 728 | 4621938 | ||
BC016971.1 | - | 6 | 748 | 3885455 | ||
BC067788.1 | - | 6 | 737 | 5298548 | ||
BF339623.1 | - | 1 | 40 | |||
BX365588.2 | - | 7 | 608 | |||
BX438861.2 | - | 6 | 771 | |||
BX460247.2 | - | 6 | 761 |