Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178450.4 | NP_848545.1 | 4 | 1068 | ||
XM_011543128.2 | XP_011541430.1 | 3 | 805 | |||
XM_011543129.1 | XP_011541431.1 | 5 | 933 | |||
XM_011543130.1 | XP_011541432.1 | 5 | 838 | |||
XM_011543131.2 | XP_011541433.1 | 5 | 1067 | |||
XM_017009014.1 | XP_016864503.1 | 3 | 1256 | |||
GenBank mRNA | AF055007.1 | - | 4 | 724 | ||
AK290129.1 | - | 4 | 852 | |||
BC047569.1 | - | 4 | 818 | 4819831 |