Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102651.1 | NP_001096121.1 | 4 | 516 | ||
XM_006722635.3 | XP_006722698.1 | 3 | 203 | |||
XM_011527660.2 | XP_011525962.1 | 4 | 357 | |||
XM_011527662.2 | XP_011525964.1 | 3 | 259 | |||
XM_011527664.2 | XP_011525966.1 | 2 | 1488 | |||
XM_011527667.2 | XP_011525969.1 | 5 | 536 | |||
XM_017026234.1 | XP_016881723.1 | 4 | 430 | |||
XM_017026235.1 | XP_016881724.1 | 2 | 1833 | |||
XM_017026236.1 | XP_016881725.1 | 2 | 201 | |||
GenBank mRNA | AK027860.1 | - | 1 | 1812 | ||
AK092136.1 | - | 5 | 568 | |||
AK299752.1 | - | 4 | 505 | |||
BC000113.1 | - | 4 | 427 | |||
BC047777.1 | - | 4 | 485 | 6187000 | ||
DC319914.1 | - | 4 | 517 |