Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321287.1 | NP_001308216.1 | 11, 12 | 1850 | ||
NM_001321288.1 | NP_001308217.1 | 10, 11 | 1171 | |||
NM_001321289.1 | NP_001308218.1 | 10, 11 | 1244 | |||
NM_001321290.1 | NP_001308219.1 | 11, 12 | 1434 | |||
NM_052936.4 | NP_443168.2 | 10, 11 | 1128 | |||
NM_178270.3 | NP_840054.1 | 9, 10 | 942 | |||
NM_178271.2 | NP_840055.1 | 11, 12 | 1430 | |||
NR_135608.1 | 9, 10 | 1135 | ||||
XM_011530841.2 | XP_011529143.1 | 9, 10 | 843 | |||
XM_011530842.1 | XP_011529144.1 | 11, 12 | 1356 | |||
GenBank mRNA | AB066214.1 | - | 9, 10 | 968 | ||
AJ320508.1 | - | 10, 11 | 986 | |||
AJ320509.1 | - | 9, 10 | 800 | |||
AJ504651.1 | - | 10, 11 | 997 | |||
AK054927.1 | - | 7, 8 | 753 | |||
AK123287.1 | - | 1, 2 | 1958 | |||
AK293533.1 | - | 10, 11 | 1123 | |||
AK314429.1 | - | 10, 11 | 995 | |||
BC041862.1 | - | 11, 12 | 1313 | 5271295 | ||
BC061696.1 | - | 10, 11 | 1130 | 4594225 |