Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024907.6 | NP_079183.4 | 6 | 987 | ||
NM_148169.2 | NP_680474.1 | 6 | 874 | |||
NR_104026.1 | 6 | 1025 | ||||
GenBank mRNA | AF386743.1 | - | 6 | 914 | ||
AK021860.1 | - | 3 | 1726 | |||
AK057934.1 | - | 6 | 874 | |||
BC012385.1 | - | 6 | 825 | 3864760 | ||
BX338290.2 | - | 6 | 914 |