Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256409.1 | NP_001243338.1 | 3 | 447 | ||
NM_052940.4 | NP_443172.1 | 2 | 413 | |||
XM_006710327.3 | XP_006710390.1 | 3 | 269 | |||
XM_006710328.3 | XP_006710391.1 | 3 | 782 | |||
GenBank mRNA | AK074521.1 | - | 2 | 386 | ||
AK075201.1 | - | 2 | 375 | |||
AK090881.1 | - | 3 | 696 | |||
AK095767.1 | - | 3 | 366 | |||
BC013101.1 | - | 2 | 252 | 3918785 | ||
CX783955.1 | - | 3 | 447 |