Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256409.1 | NP_001243338.1 | 9 | 1489 | ||
NM_052940.4 | NP_443172.1 | 8 | 1455 | |||
XM_006710327.3 | XP_006710390.1 | 9 | 1311 | |||
XM_006710328.3 | XP_006710391.1 | 9 | 1824 | |||
GenBank mRNA | AK074521.1 | - | 8 | 1428 | ||
AK075201.1 | - | 8 | 1417 | |||
AK090881.1 | - | 9 | 1738 | |||
AK095767.1 | - | 9 | 1408 | |||
BC013101.1 | - | 8 | 1294 | 3918785 |