Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052941.4 | NP_443173.2 | 2 | 317 | ||
GenBank mRNA | AF288814.1 | - | 2 | 201 | ||
AK131094.1 | - | 2 | 230 | |||
AK304245.1 | - | 2, 3 | 196 | |||
AK312417.1 | - | 2 | 309 | |||
AL832576.2 | - | 2 | 247 | |||
BC017889.1 | - | 2 | 292 | |||
BC070055.1 | - | 2 | 254 | 30345441 |