Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138446.1 | NP_612455.1 | 4 | 605 | ||
XM_017011731.1 | XP_016867220.1 | 4 | 723 | |||
GenBank mRNA | AK127742.1 | - | 4 | 585 | ||
AK316559.1 | - | 4 | 585 | |||
AL832885.1 | - | 1 | 1469 | |||
BC012331.1 | - | 4 | 605 | 4646294 |