Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152896.2 | NP_690856.1 | 12 | 2235 | ||
NR_046386.1 | 13 | 2334 | ||||
XM_011517704.2 | XP_011516006.1 | 12 | 1864 | |||
XM_011517705.2 | XP_011516007.1 | 10 | 1363 | |||
XM_017014253.1 | XP_016869742.1 | 11 | 2019 | |||
XM_017014254.1 | XP_016869743.1 | 13 | 1967 | |||
GenBank mRNA | AB071698.1 | - | 12 | 1933 | ||
AF274049.1 | - | 12 | 2063 | |||
AL833522.1 | - | 6 | 2070 | |||
BC028397.2 | - | 13 | 2334 | 4797830 |