Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138447.2 | NP_612456.1 | 3 | 965 | ||
NR_073480.1 | 3 | 600 | ||||
NR_073481.1 | 3 | 603 | ||||
NR_073482.1 | 3 | 483 | ||||
NR_104166.1 | 3 | 502 | ||||
GenBank mRNA | AB065282.1 | - | 3 | 953 | ||
AK074896.1 | - | 3 | 688 | |||
AK310459.1 | - | 3 | 522 | |||
BC014000.2 | - | 3 | 940 | 3936339 | ||
CD243730.1 | - | 1 | 150 | |||
CN406120.1 | - | 3 | 614 | |||
DB456506.1 | - | 4 | 483 |