Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258413.2 | NP_001245342.1 | 6 | 666 | ||
NM_001258414.1 | NP_001245343.1 | 6 | 666 | |||
NM_052944.4 | NP_443176.2 | 6 | 676 | |||
XM_005255080.2 | XP_005255137.1 | 7 | 704 | |||
XM_017022900.1 | XP_016878389.1 | 7 | 1020 | |||
XM_017022901.1 | XP_016878390.1 | 7 | 1730 | |||
XM_017022902.1 | XP_016878391.1 | 7 | 859 | |||
XM_017022903.1 | XP_016878392.1 | 8 | 869 | |||
XM_017022904.1 | XP_016878393.1 | 8 | 1039 | |||
XM_017022906.1 | XP_016878395.1 | 7 | 880 | |||
XM_017022907.1 | XP_016878396.1 | 7 | 704 | |||
XM_017022908.1 | XP_016878397.1 | 7 | 763 | |||
GenBank mRNA | AF292385.1 | - | 5 | 444 | ||
AJ305237.1 | - | 5 | 462 | |||
AK125267.1 | - | 8 | 792 | |||
AK298373.1 | - | 5 | 461 | |||
AY044906.1 | - | 6 | 676 | |||
BC049385.1 | - | 6 | 536 | 5755223 | ||
BC057780.1 | - | 6 | 535 | 5287366 | ||
U41898.1 | - | 3 | 228 |