Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOSTRIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039724.3 | NP_001034813.2 | 10 | 895 | ||
NM_001171631.1 | NP_001165102.1 | 15 | 1767 | |||
NM_001171632.1 | NP_001165103.1 | 9 | 811 | |||
NM_052946.3 | NP_443178.2 | 9 | 802 | |||
XM_005246270.1 | XP_005246327.1 | 10 | 954 | |||
XM_006712233.3 | XP_006712296.1 | 11 | 1017 | |||
XM_011510540.2 | XP_011508842.1 | 11 | 1017 | |||
XM_011510543.2 | XP_011508845.1 | 4 | 470 | |||
XM_017003278.1 | XP_016858767.1 | 10 | 846 | |||
XM_017003279.1 | XP_016858768.1 | 10 | 1340 | |||
XM_017003280.1 | XP_016858769.1 | 9 | 817 | |||
GenBank mRNA | AJ532842.1 | - | 10 | 838 | ||
AK002203.2 | - | 10 | 867 | |||
AK093444.1 | - | 15 | 1765 | |||
AK290254.1 | - | 10 | 861 | |||
BC014189.2 | - | 9 | 802 | 4650302 | ||
BC093072.1 | - | 10 | 842 | 30327154 | ||
DQ402497.1 | - | 9 | 754 |