Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052947.3 | NP_443179.3 | 13 | 6545 | ||
XM_011525801.2 | XP_011524103.1 | 12 | 6729 | |||
XM_011525802.2 | XP_011524104.1 | 11 | 7165 | |||
XM_017025538.1 | XP_016881027.1 | 10 | 5663 | |||
XM_017025539.1 | XP_016881028.1 | 10 | 4411 | |||
GenBank mRNA | AL832018.2 | - | 10 | 4877 | ||
AY044450.1 | - | 10 | 4612 | |||
BX647446.1 | - | 9 | 4023 | |||
BX647639.1 | - | 10 | 5411 | |||
BX647796.1 | - | 13 | 6532 |