Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT61A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152307.2 | NP_689520.2 | 2 | 379 | ||
GenBank mRNA | AK097771.1 | - | 2 | 379 | ||
BC010167.2 | - | 2 | 362 | 3960186 | ||
DB496873.1 | - | 2 | 379 |