Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT61A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152307.2 | NP_689520.2 | 4 | 1620 | ||
GenBank mRNA | AK097771.1 | - | 5 | 1121 | ||
BC007241.1 | - | 1 | 200 | |||
BC016033.2 | - | 3 | 1206 |