Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143688.2 | NP_001137160.1 | 8 | 1092 | ||
NM_001323936.1 | NP_001310865.1 | 8 | 1011 | |||
NM_001323937.1 | NP_001310866.1 | 7 | 913 | |||
NM_001323938.1 | NP_001310867.1 | 8 | 1019 | |||
NM_001323939.1 | NP_001310868.1 | 9 | 1216 | |||
NM_001323941.1 | NP_001310870.1 | 8 | 1487 | |||
NM_001323944.1 | NP_001310873.1 | 8 | 1286 | |||
NM_001323945.1 | NP_001310874.1 | 8 | 1560 | |||
NM_001323946.1 | NP_001310875.1 | 8 | 1213 | |||
NM_001323948.1 | NP_001310877.1 | 7 | 1157 | |||
NM_133375.4 | NP_588616.1 | 8 | 1042 | |||
XM_005254146.3 | XP_005254203.1 | 8 | 1160 | |||
GenBank mRNA | AB075835.1 | - | 7 | 749 | ||
AK095407.1 | - | 8 | 1071 | |||
BC022089.2 | - | 8 | 977 | 3029342 |