Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138451.2 | NP_612460.1 | 2 | 817 | ||
XM_005253832.4 | XP_005253889.1 | 2 | 1060 | |||
XM_005253833.4 | XP_005253890.1 | 2 | 1308 | |||
XM_011537862.2 | XP_011536164.1 | 2 | 1940 | |||
XM_011537863.2 | XP_011536165.1 | 3 | 1186 | |||
XM_011537864.1 | XP_011536166.1 | 2 | 817 | |||
GenBank mRNA | AK127152.1 | - | 2 | 817 | ||
BC013151.1 | - | 2 | 798 | 4385421 |