Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052950.3 | NP_443182.1 | 3 | 566 | ||
XM_011534914.1 | XP_011533216.1 | 3 | 457 | |||
GenBank mRNA | AF411978.1 | - | 3 | 226 | ||
AK309815.1 | - | 3 | 419 | |||
BC014004.2 | - | 3 | 406 | 3842589 |