Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052950.3 | NP_443182.1 | 11 | 1501 | ||
XM_011534914.1 | XP_011533216.1 | 11 | 1392 | |||
XM_011534915.2 | XP_011533217.1 | 10 | 1581 | |||
XM_017020386.1 | XP_016875875.1 | 11 | 7305 | |||
GenBank mRNA | AF411978.1 | - | 11 | 1161 | ||
BC014004.2 | - | 11 | 1341 | 3842589 |