Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317915.1 | NP_001304844.1 | 3, 4 | 575 | ||
NM_138453.3 | NP_612462.1 | 3, 4 | 537 | |||
GenBank mRNA | AY026936.1 | - | 3, 4 | 495 | ||
BC013033.2 | - | 3, 4 | 469 | 3534915 |