Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTHRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256099.1 | NP_001243028.1 | 2 | 328 | ||
NM_138455.3 | NP_612464.1 | 2 | 393 | |||
XM_011516824.2 | XP_011515126.1 | 2 | 358 | |||
GenBank mRNA | AF395488.1 | - | 2 | 329 | ||
AK130063.1 | - | 3 | 316 | |||
AK300273.1 | - | 2 | 358 | |||
AY136825.1 | - | 2 | 354 | |||
AY358914.1 | - | 2 | 388 | |||
BC014245.2 | - | 2 | 350 | 4586039 | ||
BC021025.1 | - | 2 | 393 | |||
BI763295.1 | - | 1 | 27 |