Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTHRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256099.1 | NP_001243028.1 | 2, 3 | 458 | ||
NM_138455.3 | NP_612464.1 | 2, 3 | 523 | |||
GenBank mRNA | AF395488.1 | - | 2, 3 | 459 | ||
AK130063.1 | - | 3, 4 | 446 | |||
AY136825.1 | - | 2, 3 | 484 | |||
AY358914.1 | - | 2, 3 | 518 | |||
BC014245.2 | - | 2, 3 | 480 | 4586039 | ||
BC021025.1 | - | 2, 3 | 523 | |||
BI763295.1 | - | 1, 2 | 157 |