Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001410.2 | NP_001001410.1 | 3, 4 | 594 | ||
GenBank mRNA | BC013943.1 | - | 3, 4 | 598 | 4064325 | |
BC016699.1 | - | 3, 4 | 598 | 4097703 | ||
BC017325.2 | - | 3, 4 | 578 | 4549940 |