Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302453.1 | NP_001289382.1 | 13 | 1585 | ||
NM_001302454.1 | NP_001289383.1 | 11 | 1699 | |||
NM_145045.4 | NP_659482.3 | 13 | 1879 | |||
GenBank mRNA | AK056363.1 | - | 13 | 1800 | ||
AK294355.1 | - | 15 | 1779 | |||
AK302113.1 | - | 13 | 1586 | |||
AK308072.1 | - | 11 | 1661 | |||
BC014252.2 | - | 12 | 1436 | |||
BC141828.1 | - | 13 | 1811 | 40148425 | ||
BC142637.1 | - | 13 | 1824 | 40147788 | ||
BC144421.1 | - | 11 | 1358 |