Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142462.2 | NP_001135934.1 | 2 | 1079 | ||
NM_001286841.1 | NP_001273770.1 | 3 | 1328 | |||
NM_053001.3 | NP_443727.2 | 2 | 1079 | |||
XM_005250778.4 | XP_005250835.1 | 3 | 1287 | |||
XM_011516825.2 | XP_011515127.1 | 3 | 1094 | |||
XM_011516826.2 | XP_011515128.1 | 2 | 720 | |||
XM_011516827.2 | XP_011515129.1 | 2 | 827 | |||
XM_017013018.1 | XP_016868507.1 | 2 | 699 | |||
GenBank mRNA | AK074518.1 | - | 2 | 1040 | ||
AK291491.1 | - | 2 | 1040 | |||
AK304654.1 | - | 3 | 1072 | |||
AK310994.1 | - | 2 | 1040 | |||
AY038072.1 | - | 2 | 859 | |||
AY038073.1 | - | 2 | 1041 | |||
BC016936.1 | - | 2 | 1025 | 4425397 |