Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301680.1 | NP_001288609.1 | 5 | 501 | ||
NM_001374.2 | NP_001365.1 | 5 | 524 | |||
XM_011522399.1 | XP_011520701.1 | 5 | 739 | |||
GenBank mRNA | AK098028.1 | - | 5 | 1361 | ||
AY298957.1 | - | 4 | 483 | |||
BC035205.1 | - | 5 | 499 | 4779847 | ||
BC063710.1 | - | 5 | 524 | 4800855 | ||
U62647.1 | - | 4 | 483 |