Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138459.3 | NP_612468.1 | 2 | 665 | ||
GenBank mRNA | AK125119.1 | - | 2 | 627 | ||
AK126335.1 | - | 3 | 379 | |||
BC013026.2 | - | 2 | 612 | 3531630 | ||
BC063794.1 | - | 2 | 619 | 6500832 | ||
BC066910.1 | - | 2 | 659 | 4830860 | ||
BC110325.1 | - | 2 | 621 | 5553827 | ||
BC150654.1 | - | 2 | 632 | 9057025 | ||
BC150655.1 | - | 2 | 632 | 9057026 |