Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138459.3 | NP_612468.1 | 5 | 1029 | ||
GenBank mRNA | AK125119.1 | - | 5 | 991 | ||
AK126335.1 | - | 6 | 743 | |||
AK130093.1 | - | 4 | 346 | |||
BC013026.2 | - | 5 | 976 | 3531630 | ||
BC063794.1 | - | 5 | 983 | 6500832 | ||
BC066910.1 | - | 5 | 1023 | 4830860 | ||
BC110325.1 | - | 5 | 985 | 5553827 | ||
BC150654.1 | - | 5 | 996 | 9057025 | ||
BC150655.1 | - | 5 | 996 | 9057026 |