Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320768.1 | NP_001307697.1 | 1 | 528 | ||
NM_052954.3 | NP_443186.2 | 1 | 528 | |||
NR_135472.1 | 1 | 528 | ||||
XM_011529450.2 | XP_011527752.1 | 1 | 408 | |||
GenBank mRNA | AF401639.1 | - | 1 | 133 | ||
AK054581.1 | - | 1 | 408 | |||
AK223576.1 | - | 1 | 245 | |||
AK304124.1 | - | 1 | 408 | |||
AK313231.1 | - | 1 | 314 | |||
AL833200.1 | - | 1 | 339 | |||
AY061853.1 | - | 1 | 73 | |||
BC036761.1 | - | 1 | 316 | 5588436 | ||
DC391430.1 | - | 1 | 528 | |||
JF693912.1 | - | 1 | 76 | |||
JF693913.1 | - | 1 | 133 | |||
KC843341.1 | - | 1 | 123 | |||
KF154022.1 | - | 1 | 133 |