Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204897.1 | NP_001191826.1 | 4 | 572 | ||
NM_001204898.1 | NP_001191827.1 | 3 | 494 | |||
NM_138461.3 | NP_612470.2 | 4 | 572 | |||
GenBank mRNA | AF192910.1 | - | 4 | 528 | ||
AK310209.1 | - | 4 | 542 | |||
BC146985.1 | - | 4 | 473 | 9056913 | ||
BC146993.1 | - | 4 | 473 | 9056921 | ||
BC147021.1 | - | 4 | 473 | 9056949 | ||
BC171870.1 | - | 3 | 395 | 9054524 | ||
BP395549.1 | - | 1 | 12 | |||
BP396791.1 | - | 1 | 17 | |||
BQ678650.1 | - | 4 | 538 | |||
CA489370.1 | - | 3 | 494 |
Gene Symbol: TM4SF19-TCTEX1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037950.1 | 4 | 572 |