Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138785.3 | NP_620140.1 | 4 | 363 | ||
GenBank mRNA | AK289402.1 | - | 4 | 350 | ||
AK295690.1 | - | 3 | 292 | |||
AK311061.1 | - | 2 | 190 | |||
AK315729.1 | - | 4 | 360 | |||
AY358952.1 | - | 4 | 481 | |||
BC014320.1 | - | 4 | 367 | 3686673 |