Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058165.2 | NP_477513.2 | 6 | 965 | ||
GenBank mRNA | AF384163.1 | - | 6 | 962 |