Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318890.1 | NP_001305819.1 | 11 | 1570 | ||
NM_052956.2 | NP_443188.2 | 10 | 1434 | |||
XM_006721016.2 | XP_006721079.1 | 9 | 1311 | |||
XM_006721017.2 | XP_006721080.1 | 8 | 1230 | |||
XM_006721018.2 | XP_006721081.1 | 8 | 792 | |||
XM_011545731.2 | XP_011544033.1 | 8 | 1609 | |||
XM_011545732.2 | XP_011544034.1 | 9 | 851 | |||
XM_017022915.1 | XP_016878404.1 | 9 | 2261 |