Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001376.4 | NP_001367.2 | 50 | 9827 | ||
GenBank mRNA | AB002323.3 | - | 50 | 9699 | ||
AB290157.1 | - | 50 | 9827 | |||
AL833600.1 | - | 22 | 3825 | |||
BX648670.1 | - | 15 | 2429 |