Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133445.2 | NP_597702.2 | 9 | 3809 | ||
GenBank mRNA | AB075853.1 | - | 9 | 3809 | ||
AJ416950.2 | - | 9 | 3223 | |||
AL359651.1 | - | 4 | 616 | |||
BC132866.1 | - | 9 | 3635 | 40146888 | ||
BC132868.1 | - | 9 | 3635 | 40146890 | ||
BC167432.1 | - | 9 | 3809 | 8860109 |