Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138983.2 | NP_620450.2 | 1 | 1771 | ||
GenBank mRNA | AL355743.1 | - | 1 | 915 | ||
AL834450.1 | - | 1 | 1764 | |||
BC011252.1 | - | 1 | 134 | |||
BC026989.1 | - | 1 | 1648 | 4944525 | ||
BC033290.1 | - | 1 | 1778 | 4940343 |