Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007533.3 | NP_001007534.1 | 1 | 176 | ||
GenBank mRNA | AF434846.1 | - | 1 | 154 | ||
AK096888.1 | - | 1 | 176 | |||
BC093707.1 | - | 1 | 119 | 7939552 | ||
BC112094.1 | - | 1 | 119 | 8327562 | ||
BX281734.1 | - | 1 | 159 | |||
DA894509.1 | - | 1 | 176 |